Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177320.2 | NP_796294.2 | 12 | 1996 | ||
GenBank mRNA | AK089483.1 | - | 12 | 2001 | ||
AK149847.1 | - | 12 | 1964 | |||
AK154643.1 | - | 12 | 1975 | |||
AK161050.1 | - | 3 | 904 | |||
AK171279.1 | - | 11 | 2121 | |||
AY156924.1 | - | 11 | 1845 | |||
BC128076.1 | - | 11 | 1845 | 40060132 | ||
BC128077.1 | - | 11 | 1845 | 40060133 |