Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190979.2 | NP_001177908.1 | 2 | 301 | ||
NM_001190980.2 | NP_001177909.1 | 2 | 301 | |||
NM_001320080.1 | NP_001307009.1 | 2 | 301 | |||
NM_005748.5 | NP_005739.2 | 2 | 301 | |||
NR_135139.1 | 2 | 301 | ||||
NR_135140.1 | 2 | 301 | ||||
XM_006719184.3 | XP_006719247.1 | 1 | 87 | |||
XM_011537728.2 | XP_011536030.1 | 2 | 226 | |||
XM_017018669.1 | XP_016874158.1 | 1 | 43 | |||
XM_017018670.1 | XP_016874159.1 | 2 | 239 | |||
GenBank mRNA | AK127531.1 | - | 2 | 181 | ||
AK291355.1 | - | 2 | 181 | |||
AK296191.1 | - | 2 | 181 | |||
BC037777.2 | - | 2 | 169 | 5276663 | ||
CB963485.1 | - | 2 | 156 | |||
CB993159.1 | - | 2 | 180 | |||
DA011078.1 | - | 2 | 282 | |||
DA491895.1 | - | 2 | 285 | |||
HY111766.1 | - | 2 | 301 |