Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205341.1 | NP_001192270.1 | 2 | 388 | ||
NM_177373.4 | NP_796347.2 | 2 | 388 | |||
GenBank mRNA | AK030621.1 | - | 2 | 359 | ||
AK220491.1 | - | 2 | 345 | |||
BC065133.1 | - | 2 | 359 | 30536236 | ||
EU568870.1 | - | 2 | 359 |