Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 25 | 4127 | ||
NM_001281749.1 | NP_001268678.1 | 24 | 4003 | |||
NM_025164.4 | NP_079440.3 | 24 | 4183 | |||
XM_005271481.2 | XP_005271538.1 | 25 | 4365 | |||
XM_005271482.4 | XP_005271539.2 | 24 | 4221 | |||
XM_005271484.3 | XP_005271541.2 | 24 | 4041 | |||
XM_005271485.3 | XP_005271542.1 | 12 | 2703 | |||
XM_011542722.1 | XP_011541024.1 | 25 | 4185 | |||
XM_011542723.2 | XP_011541025.1 | 25 | 4208 | |||
XM_011542724.2 | XP_011541026.1 | 24 | 4062 | |||
XM_011542725.2 | XP_011541027.1 | 23 | 3945 | |||
XM_017017424.1 | XP_016872913.1 | 23 | 3897 | |||
XM_017017425.1 | XP_016872914.1 | 25 | 4403 | |||
XM_017017426.1 | XP_016872915.1 | 23 | 4277 | |||
XM_017017427.1 | XP_016872916.1 | 20 | 3680 | |||
GenBank mRNA | AB209327.1 | - | 17 | 3193 | ||
AK022302.1 | - | 6 | 1738 | |||
AK122838.1 | - | 10 | 3445 | |||
BC063887.1 | - | 10 | 2196 | |||
BC086858.1 | - | 6 | 1738 |