Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195632.1 | NP_001182561.1 | 16 | 1641 | ||
NM_177379.4 | NP_796353.3 | 7 | 977 | |||
GenBank mRNA | AB088417.1 | - | 8 | 924 | ||
AK036860.1 | - | 2 | 1709 | |||
AY194286.1 | - | 7 | 704 | |||
BC132390.1 | - | 6 | 633 | 40130667 | ||
BC138042.1 | - | 6 | 633 | 8861061 |