Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177383.4 | NP_796357.1 | 2 | 838 | ||
GenBank mRNA | AK048682.1 | - | 2 | 839 | ||
AK139361.1 | - | 2 | 839 | |||
AK141999.1 | - | 872 | ||||
AY255598.1 | - | 1 | 148 | |||
BC112377.1 | - | 1 | 721 | 40057664 |
Set Membership: |
G-protein coupled receptor |