Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175275.4 | NP_780484.2 | 3, 4 | 695 | ||
NM_177385.4 | NP_796359.1 | 3, 4 | 695 | |||
GenBank mRNA | AK031604.1 | - | 3, 4 | 601 | ||
AK052558.1 | - | 3, 4 | 615 | |||
AK165111.1 | - | 2, 3 | 168 | |||
BC125622.1 | - | 3, 4 | 570 | |||
BC125624.1 | - | 3, 4 | 570 |