Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177390.3 | NP_796364.2 | 1, 2 | 325 | ||
GenBank mRNA | AK037051.1 | - | 1, 2 | 304 | ||
AK049374.1 | - | 1, 2 | 326 | |||
AK188302.1 | - | 1, 2 | 58 | |||
AK203977.1 | - | 1, 2 | 58 | |||
BC150765.1 | - | 1, 2 | 191 | 9087676 |