Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102474.1 | NP_001095944.1 | 4 | 673 | ||
NM_177578.4 | NP_808246.1 | 4 | 849 | |||
GenBank mRNA | AK040383.1 | - | 4 | 850 | ||
EF494894.1 | - | 4 | 673 | |||
EF494895.1 | - | 4 | 673 | |||
EF494896.1 | - | 3 | 325 |