Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177583.4 | NP_808251.1 | 6 | 2498 | ||
GenBank mRNA | AK028521.1 | - | 7 | 2757 | ||
AK049828.1 | - | 6 | 2499 | |||
AK154847.1 | - | 6 | 2566 | |||
AK162585.1 | - | 7 | 2733 | |||
BC079659.1 | - | 6 | 2530 | 6853991 |