Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPCAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005768.5 | NP_005759.4 | 5, 6 | 609 | ||
GenBank mRNA | AK096775.1 | - | 5, 6 | 798 | ||
AK296145.1 | - | 4, 5 | 319 | |||
AK311727.1 | - | 3, 4 | 331 | |||
AK315538.1 | - | 3, 4 | 181 | |||
AU125262.1 | - | 5, 6 | 681 | |||
BC000664.2 | - | 4, 5 | 344 | |||
BC065194.1 | - | 5, 6 | 540 | 5227816 | ||
BT007000.1 | - | 3, 4 | 177 | |||
BX647983.1 | - | 5, 6 | 549 | |||
BX648009.1 | - | 5, 6 | 580 | |||
U72515.1 | - | 4, 5 | 294 |