Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166395.1 | NP_001159867.1 | 2 | 925 | ||
NM_005769.2 | NP_005760.1 | 2 | 873 | |||
GenBank mRNA | AF131235.1 | - | 2 | 843 | ||
AF149783.1 | - | 1 | 754 | |||
AF280088.1 | - | 2 | 783 | |||
AK026635.1 | - | 2 | 733 | |||
AK074746.1 | - | 2 | 730 | |||
BC035282.1 | - | 2 | 739 | 5179826 |