Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam167a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177628.4 | NP_808296.2 | 4 | 2328 | ||
GenBank mRNA | AK079469.1 | - | 2 | 1404 | ||
BC060626.1 | - | 2 | 1441 | |||
BC065085.1 | - | 4 | 2111 | 6849398 | ||
BC094561.1 | - | 4 | 2238 |