Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177642.3 | NP_808310.2 | 27 | 8811 | ||
GenBank mRNA | AK036509.1 | - | 8 | 1586 | ||
EU429481.1 | - | 13 | 5683 | |||
EU429482.1 | - | 12 | 2550 | |||
GU290042.1 | - | 27 | 8811 |