Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177671.5 | NP_808339.2 | 3 | 966 | ||
GenBank mRNA | AK039700.1 | - | 3 | 929 | ||
AK143168.1 | - | 1851 | ||||
BC116991.1 | - | 3 | 896 | 40126310 | ||
BC119084.1 | - | 3 | 896 | 8733833 |