Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931409K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177676.6 | NP_808344.3 | 16, 17 | 2038 | ||
GenBank mRNA | AK029839.1 | - | 15, 16 | 2470 | ||
BC137999.1 | - | 12, 13 | 1759 | 8861018 | ||
BC145792.1 | - | 12, 13 | 1761 | 40131095 |