Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289589.1 | NP_001276518.1 | 5 | 2859 | ||
NM_001289590.1 | NP_001276519.1 | 5 | 2929 | |||
NM_177681.4 | NP_808349.1 | 4 | 2763 | |||
GenBank mRNA | AK049650.1 | - | 4 | 2719 | ||
AK136947.1 | - | 1 | 965 | |||
BC053080.1 | - | 5 | 2768 | 5702966 | ||
BC059836.1 | - | 5 | 2840 | 5697949 |