Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177690.3 | NP_808358.2 | 5 | 896 | ||
GenBank mRNA | AK054264.1 | - | 5 | 897 | ||
AY360473.1 | - | 5 | 897 | |||
DQ066881.1 | - | 5 | 897 |