Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271879.1 | NP_001258808.1 | 12 | 1365 | ||
NM_177694.6 | NP_808362.2 | 13 | 1407 | |||
GenBank mRNA | AB125740.1 | - | 13 | 1407 | ||
AB206762.1 | - | 13 | 1407 | |||
AK034197.1 | - | 13 | 1389 | |||
BC109163.2 | - | 12 | 1287 | 40047987 |