Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4932411N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177705.3 | NP_808373.1 | 3 | 1220 | ||
GenBank mRNA | AK029976.1 | - | 3 | 1220 | ||
BC117864.1 | - | 1 | 826 | 40095418 | ||
BC117865.1 | - | 1 | 826 | 40095419 |