Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata31d1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177711.3 | NP_808379.2 | 4 | 2478 | ||
GenBank mRNA | AK077018.1 | - | 4 | 2480 |