Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290395.1 | NP_001277324.1 | 7 | 1117 | ||
NM_177746.4 | NP_808414.2 | 6 | 747 | |||
GenBank mRNA | AK034920.1 | - | 7 | 1118 | ||
AY611031.1 | - | 6 | 722 | |||
AY611032.1 | - | 6 | 747 | |||
BC138085.1 | - | 6 | 715 | 8861104 | ||
BC138086.1 | - | 6 | 715 | 8861105 |