Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177755.3 | NP_808423.1 | 2 | 1056 | ||
GenBank mRNA | AK033353.1 | - | 2 | 1057 | ||
BC132483.1 | - | 2 | 958 | 40130760 | ||
BC145690.1 | - | 2 | 958 | 40130993 |