Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177809.4 | NP_808477.2 | 4, 5 | 564 | ||
GenBank mRNA | AK054181.1 | - | 4, 5 | 565 | ||
BC107250.1 | - | 4, 5 | 528 | 40052351 | ||
BC107251.1 | - | 4, 5 | 528 | 40052352 | ||
DV649007.1 | - | 1, 2 | 175 |