Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam135b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177819.3 | NP_808487.2 | 5 | 533 | ||
GenBank mRNA | AK043571.1 | - | 5 | 534 | ||
BC125645.1 | - | 5 | 408 | |||
BC125647.1 | - | 5 | 408 |