Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 5 | 551 | ||
NM_005817.4 | NP_005808.3 | 5 | 551 | |||
GenBank mRNA | AF055574.1 | - | 5 | 427 | ||
AF057140.1 | - | 5 | 448 | |||
AK223054.1 | - | 5 | 481 | |||
AK225045.1 | - | 5 | 417 | |||
AK225400.1 | - | 5 | 432 | |||
AK226088.1 | - | 5 | 429 | |||
AK291104.1 | - | 5 | 508 | |||
AK312740.1 | - | 5 | 477 | |||
BC001590.2 | - | 5 | 424 | 2987965 | ||
BC005818.2 | - | 5 | 440 | 3833411 | ||
BC007566.1 | - | 5 | 440 | 3028104 | ||
BC019278.1 | - | 5 | 440 | 2905275 | ||
BF683925.1 | - | 5 | 425 | |||
BG339642.1 | - | 5 | 427 | |||
BQ057849.1 | - | 5 | 438 | |||
BQ063091.1 | - | 4 | 405 | |||
BQ276395.1 | - | 5 | 438 | |||
BT007235.1 | - | 4 | 374 | |||
DB127412.1 | - | 5 | 508 |