Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A530013C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040674.1 | - | 2298 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177851.1 | - | 2298 | Chr. - - [-] on Build |