Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278784.1 | NP_001265713.1 | 1, 2 | 222 | ||
NM_005827.2 | NP_005818.2 | 1, 2 | 222 | |||
GenBank mRNA | AK293563.1 | - | 1, 2 | 222 | ||
AK296073.1 | - | 1, 2 | 191 | |||
AU099388.1 | - | 1, 2 | 226 | |||
AU309730.1 | - | 1, 2 | 256 | |||
BC011888.2 | - | 1, 2 | 201 | 3832062 | ||
D87989.1 | - | 1, 2 | 191 |