Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177876.4 | NP_808544.1 | 4 | 816 | ||
GenBank mRNA | AK082658.1 | - | 4 | 782 | ||
AK149893.1 | - | 4 | 783 | |||
AK166359.1 | - | 4 | 797 | |||
AK172114.1 | - | 4 | 776 | |||
AK172337.1 | - | 4 | 779 | |||
AK205523.1 | - | 1 | 62 | |||
BC026744.1 | - | 4 | 817 | 4163734 |