Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177876.4 | NP_808544.1 | 4 | 1026 | ||
GenBank mRNA | AK082658.1 | - | 4 | 992 | ||
AK149893.1 | - | 4 | 993 | |||
AK166359.1 | - | 4 | 1007 | |||
AK172114.1 | - | 4 | 986 | |||
AK172337.1 | - | 4 | 990 | |||
BC026744.1 | - | 4 | 1027 | 4163734 |