Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177879.5 | NP_808547.3 | 34 | 5248 | ||
GenBank mRNA | AK032883.1 | - | 9 | 1148 | ||
AK147578.1 | - | 34 | 5293 | |||
AY353236.1 | - | 30 | 4398 | |||
BC060237.1 | - | 34 | 4832 | |||
BC138453.1 | - | 30 | 4269 | 8861473 |