Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033929.2 | NP_001029101.1 | 7, 8 | 1172 | ||
NM_178413.5 | NP_848500.3 | 8, 9 | 1240 | |||
GenBank mRNA | AI156509.1 | - | 1, 2 | 35 | ||
AK165463.1 | - | 7, 8 | 1138 | |||
BC025604.1 | - | 8, 9 | 1203 | 5250319 | ||
BC051244.1 | - | 7, 8 | 1134 | 4975661 | ||
BQ042728.1 | - | 4, 5 | 521 |