Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005847.4 | NP_005838.3 | 12, 13 | 1558 | ||
NM_152685.3 | NP_689898.2 | 12, 13 | 1570 | |||
XM_005272148.3 | XP_005272205.3 | 13, 14 | 1695 | |||
XM_005272149.3 | XP_005272206.3 | 13, 14 | 1683 | |||
XM_011543765.2 | XP_011542067.1 | 13, 14 | 1695 | |||
XM_011543766.1 | XP_011542068.1 | 12, 13 | 1476 | |||
XM_011543767.1 | XP_011542069.1 | 11, 12 | 1380 | |||
XM_011543768.1 | XP_011542070.1 | 12, 13 | 1600 | |||
XM_011543769.1 | XP_011542071.1 | 11, 12 | 1355 | |||
GenBank mRNA | AF058317.1 | - | 12, 13 | 1461 | ||
AF098277.1 | - | 12, 13 | 1508 | |||
AF170911.1 | - | 12, 13 | 1492 | |||
AJ250807.1 | - | 12, 13 | 1531 | |||
AJ269477.1 | - | 12, 13 | 1461 | |||
AK092655.1 | - | 11, 12 | 1382 | |||
AK290789.1 | - | 12, 13 | 1491 | |||
BC019225.1 | - | 5, 6 | 506 | |||
BC050261.1 | - | 12, 13 | 1512 | 5188430 |