Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178934.4 | NP_849265.2 | 2 | 318 | ||
GenBank mRNA | AJ549317.2 | - | 2 | 318 | ||
AK028970.1 | - | 2 | 291 | |||
AK031659.1 | - | 2 | 236 | |||
AK035972.1 | - | 2 | 199 | |||
AK143263.1 | - | 2 | 307 | |||
AY230881.1 | - | 2 | 217 | |||
BC096454.1 | - | 2 | 249 | 4223231 | ||
BC116314.1 | - | 2 | 179 | 40095492 | ||
BC116315.1 | - | 2 | 182 | 40095493 | ||
BC138316.1 | - | 2 | 165 | 8861336 | ||
BC138317.1 | - | 2 | 165 | 8861337 |