Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290776.1 | NP_001277705.1 | 3 | 341 | ||
NM_001290777.1 | NP_001277706.1 | 3 | 341 | |||
NM_001290778.1 | NP_001277707.1 | 2 | 488 | |||
NM_178935.5 | NP_849266.1 | 2 | 488 | |||
GenBank mRNA | AK030100.1 | - | 2 | 435 | ||
AK031023.1 | - | 1 | 1160 | |||
AK031783.1 | - | 2 | 402 | |||
AK044130.1 | - | 2 | 437 | |||
AK078477.1 | - | 3 | 288 | |||
AK134415.1 | - | 2 | 1164 | |||
AK161561.1 | - | 3 | 250 | |||
AK162263.1 | - | 2 | 352 | |||
AK166902.1 | - | 2 | 1013 | |||
BC059099.1 | - | 2 | 418 | |||
BC080778.1 | - | 2 | 400 | |||
BC117898.1 | - | 3 | 282 | 40096960 | ||
BC117899.1 | - | 3 | 282 | 40096967 | ||
DQ316984.1 | - | 2 | 488 | |||
DQ320011.1 | - | 3 | 341 |