Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005863.4 | NP_005854.2 | 1, 2 | 316 | ||
NR_073040.1 | 1, 2 | 316 | ||||
GenBank mRNA | AJ010046.1 | - | 1, 2 | 249 | ||
AK024919.1 | - | 1, 2 | 256 | |||
AK294244.1 | - | 1, 2 | 258 | |||
AK304028.1 | - | 1, 2 | 258 | |||
AK315566.1 | - | 1, 2 | 258 | |||
BC010285.1 | - | 1, 2 | 247 | 3048980 | ||
DA727151.1 | - | 1, 2 | 320 | |||
DB177813.1 | - | 1, 2 | 284 |