Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd59b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181858.1 | NP_862906.1 | 396 | |||
GenBank mRNA | BC139203.1 | - | 5 | 443 | 8862225 | |
BC145912.1 | - | 5 | 443 | 40131215 | ||
BC171969.1 | - | 5 | 443 | 9054255 |