Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr16c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181989.1 | NP_871789.1 | 5, 6 | 934 | ||
GenBank mRNA | AK136508.1 | - | 4, 5 | 905 | ||
AY277588.1 | - | 5, 6 | 934 | |||
BC104134.1 | - | 5, 6 | 853 | 40045268 | ||
BC104135.1 | - | 5, 6 | 853 | 40045269 |