Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr16c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181989.1 | NP_871789.1 | 6 | 1023 | ||
GenBank mRNA | AK136508.1 | - | 5 | 994 | ||
AY277588.1 | - | 6 | 1023 | |||
BC104134.1 | - | 6 | 942 | 40045268 | ||
BC104135.1 | - | 6 | 942 | 40045269 |