Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183158.3 | NP_898981.2 | 7 | 978 | ||
GenBank mRNA | AK089408.1 | - | 7 | 981 | ||
BC138088.1 | - | 3 | 497 | 8861107 | ||
BC145141.1 | - | 3 | 497 |