Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homez | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177705.1 | NP_001171176.1 | 2 | 702 | ||
NM_183174.3 | NP_898997.2 | 2 | 547 | |||
GenBank mRNA | AK129361.1 | - | 2 | 592 | ||
AK178386.1 | - | 1 | 53 | |||
AY258064.1 | - | 2 | 650 | |||
BC094669.1 | - | 2 | 547 | 30616315 |