Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005886.2 | NP_005877.2 | 5, 6 | 757 | ||
XM_005255772.4 | XP_005255829.1 | 5, 6 | 787 | |||
XM_006721121.3 | XP_006721184.1 | 5, 6 | 788 | |||
XM_006721122.2 | XP_006721185.1 | 5, 6 | 912 | |||
XM_006721123.3 | XP_006721186.1 | 5, 6 | 787 | |||
XM_011522810.2 | XP_011521112.1 | 5, 6 | 1426 | |||
XM_017022860.1 | XP_016878349.1 | 5, 6 | 729 | |||
XM_017022861.1 | XP_016878350.1 | 5, 6 | 912 | |||
XM_017022862.1 | XP_016878351.1 | 5, 6 | 1426 | |||
XM_017022863.1 | XP_016878352.1 | 5, 6 | 926 | |||
XM_017022864.1 | XP_016878353.1 | 5, 6 | 1426 | |||
GenBank mRNA | AB209250.1 | - | 5, 6 | 896 | ||
AF052432.1 | - | 4, 5 | 423 | |||
BC001353.2 | - | 4, 5 | 406 | 2964632 | ||
BT007022.1 | - | 4, 5 | 383 | |||
CR456762.1 | - | 4, 5 | 383 |