Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 14 | 6932 | ||
NM_001110229.1 | NP_001103699.1 | 15 | 7031 | |||
NM_001110230.1 | NP_001103700.1 | 15 | 7019 | |||
NM_001110231.1 | NP_001103701.1 | 16 | 8378 | |||
NM_001110232.1 | NP_001103702.1 | 16 | 7005 | |||
NM_001160292.1 | NP_001153764.1 | 14 | 6875 | |||
NM_001160293.1 | NP_001153765.1 | 14 | 8036 | |||
NM_010160.2 | NP_034290.2 | 12 | 8154 | |||
GenBank mRNA | AK045654.1 | - | 1 | 1071 | ||
BC033049.1 | - | 1 | 168 |