Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000162.3 | NP_000153.1 | 10 | 2167 | ||
NM_033507.1 | NP_277042.1 | 10 | 1868 | |||
NM_033508.1 | NP_277043.1 | 11 | 1992 | |||
GenBank mRNA | AK122876.1 | - | 10 | 1862 | ||
BC001890.1 | - | 10 | 2167 | 3536582 | ||
CD251038.1 | - | 1 | 130 | |||
M69051.1 | - | 11 | 1979 | |||
M88011.1 | - | 10 | 2026 | |||
M90299.1 | - | 10 | 2155 |