Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005892.3 | NP_005883.2 | 16, 17 | 2218 | ||
XM_006722062.3 | XP_006722125.1 | 17, 18 | 2433 | |||
XM_006722063.3 | XP_006722126.1 | 16, 17 | 2413 | |||
XM_006722064.3 | XP_006722127.1 | 17, 18 | 2403 | |||
XM_006722065.3 | XP_006722128.1 | 16, 17 | 2383 | |||
XM_006722066.3 | XP_006722129.1 | 17, 18 | 2429 | |||
XM_006722069.2 | XP_006722132.1 | 17, 18 | 2432 | |||
XM_006722070.3 | XP_006722133.1 | 16, 17 | 2409 | |||
XM_011525179.2 | XP_011523481.1 | 17, 18 | 2433 | |||
XM_011525180.2 | XP_011523482.1 | 17, 18 | 2125 | |||
XM_011525182.2 | XP_011523484.1 | 17, 18 | 2430 | |||
GenBank mRNA | AF432213.1 | - | 13, 14 | 1692 | ||
AJ008112.1 | - | 1, 2 | 134 | |||
AJ008115.1 | - | 1 | 58 | |||
AK131091.1 | - | 8, 9 | 2755 | |||
AY278319.1 | - | 16, 17 | 2218 | |||
BC001710.1 | - | 1, 2 | 122 | |||
BC021906.1 | - | 4, 5 | 726 | |||
BC073988.1 | - | 4, 5 | 766 | |||
BC111564.1 | - | 9, 10 | 1031 | 40008718 | ||
CR456759.1 | - | 1, 2 | 95 | |||
FJ534522.1 | - | 1 | 17 |