Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183308.2 | NP_899131.1 | 4, 5 | 462 | ||
GenBank mRNA | AK007350.1 | - | 4, 5 | 430 | ||
AK146311.1 | - | 4, 5 | 430 | |||
AK152351.1 | - | 4, 5 | 413 | |||
AK171926.1 | - | 4, 5 | 420 | |||
BC021887.1 | - | 4, 5 | 419 | 3154583 | ||
BC037140.1 | - | 4, 5 | 404 | 5027719 | ||
BC055896.1 | - | 4, 5 | 462 | 3482584 | ||
BC062200.1 | - | 4, 5 | 453 | 6399484 | ||
L48514.1 | - | 4, 5 | 398 |