Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146311.1 | - | 6 | 716 | ||
AK171926.1 | - | 6 | 706 | |||
BC037140.1 | - | 6 | 690 | 5027719 | ||
BC055896.1 | - | 6 | 748 | 3482584 | ||
BC062200.1 | - | 6 | 739 | 6399484 |