Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005754.2 | NP_005745.1 | 4 | 488 | ||
NM_198395.1 | NP_938405.1 | 4 | 457 | |||
XM_006714749.3 | XP_006714812.1 | 4 | 477 | |||
XM_006714750.3 | XP_006714813.1 | 4 | 416 | |||
XM_017008922.1 | XP_016864411.1 | 4 | 763 | |||
XM_017008923.1 | XP_016864412.1 | 4 | 732 | |||
GenBank mRNA | AK222606.1 | - | 4 | 449 | ||
AK300098.1 | - | 4 | 449 | |||
AK303580.1 | - | 4 | 479 | |||
AK309172.1 | - | 4 | 451 | |||
AK312572.1 | - | 4 | 451 | |||
BC006997.1 | - | 4 | 451 | 3950738 | ||
BC108278.1 | - | 4 | 396 | 6051310 | ||
BG703413.1 | - | 4 | 460 | |||
BT019384.1 | - | 3 | 317 | |||
BU517281.1 | - | 4 | 481 | |||
BX647869.1 | - | 4 | 450 | |||
BX648012.1 | - | 4 | 463 | |||
CR536535.1 | - | 3 | 317 | |||
U32519.1 | - | 4 | 438 |