Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198113.2 | NP_932781.1 | 7 | 706 | ||
GenBank mRNA | AB099289.1 | - | 7 | 704 | ||
AK038981.1 | - | 6 | 1278 | |||
AK148293.1 | - | 7 | 699 | |||
AK149880.1 | - | 7 | 715 | |||
AK164662.1 | - | 6 | 404 | |||
AK166342.1 | - | 7 | 712 | |||
AK171514.1 | - | 2 | 2763 | |||
BC028922.1 | - | 7 | 706 | 3987714 |