Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dagla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198114.2 | NP_932782.2 | 14 | 1490 | ||
GenBank mRNA | AK147384.1 | - | 14 | 1448 | ||
AY275376.1 | - | 13 | 1390 | |||
BC112414.1 | - | 13 | 1389 | |||
BC148308.2 | - | 13 | 1390 | 40061146 |